Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EVI2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566054 Supplier Invitrogen™ Supplier No. PA566054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLLRSWFGNKD FQALPILARL PSMPTDMEHT GHYLHLAFLM TTVFSLSPGT KANYT Highest antigen sequence identity to the following orthologs - mouse 40%, rat 46%.

May complex with itself or/and other proteins within the membrane, to function as part of a cell-surface receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EVI2A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Evi2a
Gene Accession No. P22794
Gene Alias AW491894; ecotropic viral integration site 2a; ecotropic viral integration site 2A protein; Ecotropic viral integration site 2A protein homolog; EV12A protein; EVDA; EVI2; Evi-2; EVI2A; Evi-2a; Protein EVI2A
Gene Symbols Evi2a
Host Species Rabbit
Immunogen Recombinant Human EVI2A. Recombinant protein control fragment (Product #RP-106747).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2123
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.