Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EVI2B Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299056PCP
Description
EVI2B Polyclonal antibody specifically detects EVI2B in Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
EVI2B | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
CD361 antigen, D17S376, ecotropic viral integration site 2B, Ecotropic viral integration site 2B protein homolog, EVDBprotein EVI2B, EVI-2B | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse EVI2B. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
2124 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction