Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EVI2B Monoclonal Antibody (8F3D1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548453
Description
EVI2B Monoclonal Antibody for Western Blot, ICC/IF, IHC, Flow, ELISA
CD361, also known as EVI2B (Ecotropic Viral Integration site 2B) or EVDB, is a poorly characterized type I transmembrane protein, expressed from one of three genes embedded in intron 27b of the neurofibromatosis type 1 (NF1) gene. The DNA strand that is transcribed to produce CD361 is the complementary one to the strand encoding NF1. Murine homolog to human CD361 is associated with ecotropic viral insertions, which have been implicated in the expression of murine myeloid leukemias. CD361 has been also reported to be involved in melanocyte and keratinocyte differentiation. However, it is expressed mainly in peripheral blood and bone marrow.
Specifications
EVI2B | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P34910 | |
EVI2B | |
Purified recombinant fragment of human CD361 (AA: extra 22-202) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry | |
8F3D1 | |
Unconjugated | |
EVI2B | |
CD361; D17S376; ecotropic viral integration site 2B; ecotropic viral integration site 2B protein; Ecotropic viral integration site 2B protein homolog; EVDB; Evi2; Evi-2; EVI2B; EVI-2B; Protein EVI2B; protein EVI2B-like | |
Mouse | |
Protein G | |
RUO | |
2124 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction