Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EVI2B Monoclonal Antibody (MEM-216)
GREENER_CHOICE

Catalog No. PIMA110212
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA110212 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA110212 Supplier Invitrogen™ Supplier No. MA110212
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope of CD361 / EVI2B, almost uncharacterized type I transmembrane protein with broad leukocyte expression, mostly in myeloid and B cells.

CD361, also known as EVI2B (Ecotropic Viral Integration site 2B) or EVDB, is a poorly characterized type I transmembrane protein, expressed from one of three genes embedded in intron 27b of the neurofibromatosis type 1 (NF1) gene. The DNA strand that is transcribed to produce CD361 is the complementary one to the strand encoding NF1. Murine homolog to human CD361 is associated with ecotropic viral insertions, which have been implicated in the expression of murine myeloid leukemias. CD361 has been also reported to be involved in melanocyte and keratinocyte differentiation. However, it is expressed mainly in peripheral blood and bone marrow.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EVI2B
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone MEM-216
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene EVI2B
Gene Accession No. P34910
Gene Alias CD361; D17S376; ecotropic viral integration site 2B; ecotropic viral integration site 2B protein; Ecotropic viral integration site 2B protein homolog; EVDB; Evi2; Evi-2; EVI2B; EVI-2B; Protein EVI2B; protein EVI2B-like
Gene Symbols EVI2B
Host Species Mouse
Immunogen Raji cells.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2124
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.