Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EVI5 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040620B
Description
EVI5 Polyclonal antibody specifically detects EVI5 in Human samples. It is validated for Western BlotSpecifications
EVI5 | |
Polyclonal | |
Western Blot | |
dJ846D11.1 (ecotropic viral integration site 5), ecotropic viral integration site 5, ecotropic viral integration site 5 protein homolog, EVI-5, NB4SNeuroblastoma stage 4S gene protein | |
The immunogen recognized by this antibody maps to a region between residue 525 and 575 of human Ecotropic Viral Integrations Site 5 using the numbering given in entry NP_005656.4 (GeneID 7813). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
7813 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction