Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EVI5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565818
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565818 Supplier Invitrogen™ Supplier No. PA565818
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QPPFDGIHIVN HLIGDDESFH SSDEDFIDNS LQETGVGFPL HGKSGSMSLD PAVADGSESE TEDSVLETRE SNQVVQKERP PRRRESYSTT V Highest antigen sequence identity to the following orthologs - mouse 71%, rat 72%.

EVI5 is a novel centrosomal protein with a complex expression pattern and subcellular localization, possibly involved in centrosome stability and dynamics. It may act as an oncogene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EVI5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EVI5
Gene Accession No. O60447
Gene Alias dJ846D11.1 (ecotropic viral integration site 5); ecotropic viral integration site 5; ecotropic viral integration site 5 protein; ecotropic viral integration site 5 protein homolog; EVI5; EVI-5; NB4S; Neuroblastoma stage 4S gene protein
Gene Symbols EVI5
Host Species Rabbit
Immunogen Recombinant Human EVI5. Recombinant protein control fragment (Product #RP-106490).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7813
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.