Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EVI5L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565363
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565363 Supplier Invitrogen™ Supplier No. PA565363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRKLVVGELQD ELMSVRLREA QALAEGRELR QRVVELETQD HIHRNLLNRV EA Highest antigen sequence identity to the following orthologs - mouse 91%, rat 89%.

Functions as a GTPase-activating protein (GAP) with a broad specificity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EVI5L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EVI5L
Gene Accession No. Q96CN4
Gene Alias ecotropic viral integration site 5 like; ecotropic viral integration site 5-like protein; EVI5L; EVI5-like protein
Gene Symbols EVI5L
Host Species Rabbit
Immunogen Recombinant Human EVI5L. Recombinant protein control fragment (Product #RP-104774).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115704
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.