Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EVI5L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560238
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560238 Supplier Invitrogen™ Supplier No. PA560238
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVRQQCSSAA EDLQKAQSTI RQLQEQQENP RLTEDFVSHL ETELEQSRLR ETETL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 51%.

Functions as a GTPase-activating protein (GAP) with a broad specificity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EVI5L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EVI5L
Gene Accession No. Q96CN4
Gene Alias ecotropic viral integration site 5 like; ecotropic viral integration site 5-like protein; EVI5L; EVI5-like protein
Gene Symbols EVI5L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EVI5L. Recombinant protein control fragment (Product #RP-99125).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115704
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.