Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EVL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553864
Description
Immunogen sequence: SSQRQVQNGP SPDEMDIQRR QVMEQHQQQR QESLERRTSA TGPILPPGHP SSAASAPVSC SGPPPPPPPP VPPPPTGATP PP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
Ena/VASP proteins are actin-associated proteins involved in a range of processes dependent on cytoskeleton remodeling and cell polarity such as axon guidance and lamellipodial and filopodial dynamics in migrating cells. EVL enhances actin nucleation and polymerization.
Specifications
EVL | |
Polyclonal | |
Unconjugated | |
EVL | |
AI528774; ena/vasodilator-stimulated phosphoprotein-like; Ena/VASP-like protein; Enah/Vasp-like; Ena-vasodilator stimulated phosphoprotein; Evl; Rnb6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51466 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UI08 | |
EVL | |
Recombinant protein corresponding to Human EVL. Recombinant protein control fragment (Product #RP-92389). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction