Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Evolution™ One/One Plus UV-Vis Spectrophotometers

Catalog No. 14385503
Click to view available options
Type:
Evolution One Plus UV-Vis Spec
Evolution One US Security Bundle
Evolution One US USP/EP Standards Bundle
Evolution One UV-Vis Spec
Evolution OnePlus US Security Bundle
Evolution OnePlus US USP/EP Standards Bundle
Spectral Bandwidth:
1 nm
1.0, 2.0 nm Variable

Thermo Scientific™ Evolution™ UV-Vis Spectrophotometers combine reliable and versatile hardware with easy-to-learn, easy-to-use Thermo Scientific™ Insight™ Pro Software. Together they will meet the diverse needs your organization requires from an analytical tool.

For routine, complex research and advanced applications, including current compliance, the Thermo Scientific™ Evolution™ One/One Plus UV-Vis Spectrophotometer series brings together reliable, versatile hardware, a broad range of accessories, with easy-to-use, easy-to-learn Thermo Scientific™ Insight™ Pro Software.

Webinar: Identification of protein aggregation through UV-Vis spectroscopy

For routine to research analysis, this instrument series offers real innovations that provide enhanced usability and performance - without added complexity. With straightforward software, cutting-edge instrumentation and a wide selection of accessories, the Evolution One and One Plus UV-Vis Spectrophometers work with you to perform experiments the way you want - for the results you need.

Optimized Engineering Delivers Ultimate Performance:
  • Double-beam geometry with fixed 1nm or variable spectral bandwidth options
  • Application Focused Beam Geometry (AFBG) technology enables the best performance from microcells, solid samples, and fiber optic probes (Evolution One Plus Only)
  • Xenon flash lamp requires no warm-up time allowing instant measurements
  • Meet chemical, materials science, or biological sampling and measurement needs with a broad range of accessories
Insight Pro Software—Easy-to-learn, easy-to-use
  • Reduce training time: Optimized user interface with prompting software defines clear step-by-step workflow and is easy to teach and learn
  • Prevent costly sampling and measurement errors: Create dedicated workflows for complex assay and transform multi-step workflows into one simplified, easy-to-follow method suitable for any level of user with Customized User Environment (CUE)
  • Always current: Whether it is user inputs or application requirements, or new regulations, Insight Pro Software is continuously improving with quick upgrades to deliver greater capabilities and efficiencies for your lab’s needs
Accessories for all your Sampling Needs:
  • Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete your analysis
  • Choice of Peltier Single Cell Holder or a Smart 8-Cell Peltier system for temperature control and sample monitoring from 0° to 100 °C
  • Easy software integration gives you complete control of your measurements

Regulatory/Quality Control Environments:
To use the EvolutionOne/One Plus UV-Vis Spectrophotometers in a regulated or quality control environment, we offer a wide selection of compliance products and services to reduce implementation time and assure system performance.

Evolution One/One Plus Security and Validator Bundles include:
  • Evolution One or One Plus UV-Vis Spectrometer
  • INSIGHT Pro with Security Software
  • Validator for Insight Pro US Ltr or INTL A4 Paper Version
  • Qualification Filter Kit for UV-Vis
Evolution One/One Plus USP/EP Standards Pharma Bundles include:
  • Evolution One or One Plus Spectrometer
  • INSIGHT Pro with Security Software
  • Validator for Insight Pro US Ltr or INTL A4 Paper Version
  • USP and EP UV Standards Set
  • USP and EP Visible Standards Set
TRUSTED_SUSTAINABILITY

Specifications

Baseline Flatness ±0.001 A, 200–800 nm, 1.0 nm SBW, smoothed
Detector Type Dual Silicon Photodiodes
Dimensions (L x W x H) 593 x 475 x 266 mm (23.3 x 18.7 x 10.6 in.)
Light Source Xenon Flash
Product Line Evolution One Plus
Spectral Bandwidth 1.0, 2.0 nm Variable
Type Evolution One Plus UV-Vis Spec
Wavelength Range 190 to 1100 nm
Drift <0.0005 A/hr., 500 nm, 1.0 nm SBW, 1 hr. warmup
Electrical Requirements 100/240 V, 50/60 Hz selected automatically, 150 W maximum
Includes Evolution One Plus UV-Vis Spectrophotometer
Lamp Life >5 years or longer if not using live signal
Noise
0A: ≤0.00015 A
1A: ≤0.00025 A
2A: ≤0.00050 A
260 nm, 1.0 nm SBW, RMS
Optical Design AFBG Microcell optimized; AFBG Fiber optic optimized; AFBG Materials optimized
Pharmacopoeia Compliance Testing Photometric Accuracy (60mg/L K2Cr2O7): ±0.010A
Stray Light: ≤1%T at 198 nm: KCI; ≤0.05%AT at 220 nm: Nal, Kl
Wavelength Accuracy: ±.5 nm 541.9, 546.1nm Hg emission lines, ±0.8 nm full range
Wavelength Repeatability: ≤0.05 nm, repetitive scanning of 546.1 nm Hg emission line
Photometric Accuracy 1 A: ±0.004 A, 2 A: ±0.008 A, Measured at 440 nm using neutral density filters traceable to NIST
Photometric Display -0.3 to 4.0 A
Photometric Range >3.5 A
Photometric Repeatability 1 A: ±0.0002 A
Scan Ordinate Modes Absorbance, % Transmittance, % Reflectance, Kubelka-Munk, log (1/R), log (Abs), Abs*Factor, Intensity
Scan Speed <1 to 6000 nm/min. (Variable)
Warranty Instrument (2 years), Lamp (3 years)
Wattage 150 W max.
Wavelength Accuracy ±0.5 nm (541.9, 546.1 nm mercury lines), ±0.8 nm (full range 190 to 1100 nm)
Wavelength Data Interval 10, 5, 2, 1.0, 0.5, 0.2, 0.1 nm
Wavelength Repeatability ≤0.05 nm (546.1 nm mercury line, SD of 10 measurements)
Weight (English) 32 lb.
Weight (Metric) 14.5 kg
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.