Learn More
Invitrogen™ EVOS™ Light Cube, AO 2.0
Catalog No. 12563691
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
12-563-691 1 each
1 options
Catalog No. 12-563-691 Supplier Invitrogen™ Supplier No. AMEP4970
Add to Cart
Add to Cart
Request a Quote
EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality. At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through.
  • Second-generation EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality
  • At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through
  • Like the original design, they combine condensing optics and hard-coated filters into one light engine that delivers digitally controlled fluorophore excitation
  • These LED light cubes are backwards compatible with older EVOS and Countess systems. and can be used with first-generation light cubes
  • Like the original light cubes, these LED cubes are user-changeable and automatically recognized by the instrument
  • The EVOS Light Cube, AO 2.0, is ideal for use with acridine orange, Alexa Fluor 430, Di-8 ANEPPS, DiA, and other fluorophores with excitation and emission maxima near 470 and 488 nm
  • The LED bulbs on the EVOS systems are rated for >50,000 hours, compared to 300 hours for a typical mercury bulb and 1,500 hours for a metal halide bulb
EVOS LED light cubes offer these important advantages:
  • Uniform illumination across the entire field of view for superb image quality
  • Optimized spectral compatibility to maximize true signal detection, while minimizing bleed-through for the best possible data quality
  • A short light path that affords superior detection of fluorescent signals
  • Stable illumination intensity over 50,000 hours that helps experimental reproducibility
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 each
For Research Use Only. Not for use in diagnostic procedures.