Learn More
Invitrogen™ EVOS™ Onstage Incubator (OSI-2) Perfusion Tubing Connector Kit
Description
The EVOS Onstage Incubator (OSI-2) Perfusion Tubing Connector Kit contains two flangeless fittings designed to securely connect perfusion tubing to the integrated ports on the OSI-2.
Specifications
Specifications
| Type | Tubing Connector Kit |
| Product Line | EVOS |
| Description | Perfusion accessory for OSI-2 |
| For Use With (Application) | Time-lapse imaging |
| For Use With (Equipment) | EVOS™ Onstage Incubator (OSI-2) |
| Content And Storage | Two flangeless fittings |
| Quantity | 1 Kit |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.