Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EVPL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587715
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587715 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587715 Supplier Invitrogen™ Supplier No. PA587715
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TKPSSSLSIG SIISKSPLAS PAPQSTSFFS PSFSLGLGDD SFPIAGIYDT TTDNKCSIKT AVAKNMLDPI TGQKLLEAQA ATGGIVDLLS RERYSVHKAM ERGLIENTST QRLLNAQKAF TGIEDPVTKK RLSVGEAVQK GWMPRESVLP HLQVQHLTGG LIDPKRTGRI PIQQALLSGM ISEELAQLLQ DESSYEKDLT DPISKERLSY KEAMGRCRKD PLSGLLLLPA ALEGYRCYRS ASPTVPRSLR; Positive Samples: HeLa, Mouse lung, Rat lung; Cellular Location: Cell junction, Cornified envelope, Cytoplasm, cytoskeleton, desmosome.

This gene encodes a member of the DEAH-containing family of RNA helicases. The encoded protein is an enzyme that catalyzes the ATP-dependent unwinding of double-stranded RNA and DNA-RNA complexes. This protein localizes to both the nucleus and the cytoplasm and functions as a transcriptional regulator. This protein may also be involved in the expression and nuclear export of retroviral RNAs. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 11 and 13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EVPL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EVPL
Gene Accession No. Q92817, Q9D952
Gene Alias 210 kDa cornified envelope precursor protein; 210 kDa paraneoplastic pemphigus antigen; 210-kD protein; 210kDa protein; envoplakin; EVPK; Evpl; p210
Gene Symbols EVPL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1784-2033 of human EVPL (NP_0019792).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14027, 2125, 303687
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.