Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EVX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565925
Description
Immunogen sequence: ESRKDMVVFLD GGQLGTLVGK RVSNLSEAVG SPLPEPPEKM VPRGCLSPRA VPPATRERGG GGPEEEPVDG LAGS Highest antigen sequence identity to the following orthologs - mouse 91%, rat 89%.
EVX1 is a member of the vertebrate eve-related homeo box family. This protein acts as a transcriptional repressor. A similar protein in mice is critical for early embryogenesis.
Specifications
| EVX1 | |
| Polyclonal | |
| Unconjugated | |
| EVX1 | |
| eve, even-skipped homeo box homolog 1; eve, even-skipped homeobox homolog 1; even skipped homeotic gene 1 homolog; even-skipped homeo box 1 (homolog of Drosophila); even-skipped homeobox 1; Evx1; EVX-1; Homeobox even-skipped homolog protein 1; NEWGENE_1565788 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2128 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49640 | |
| EVX1 | |
| Recombinant Human EVX1. Recombinant protein control fragment (Product #RP-106604). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction