Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Evx2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568572
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568572 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568572 Supplier Thermo Scientific Supplier No. PA568572
Add to Cart
Add to Cart

Evx2 Polyclonal Antibody

Evx2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Evx2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EVX2
Gene Accession No. 0
Gene Alias eve, even-skipped homeo box homolog 2; even skipped homeotic gene 2; even skipped homeotic gene 2 homolog; even-skipped homeo box 2 (homolog of Drosophila eve); even-skipped homeobox 2; Evx2; Evx-2; Homeobox even-skipped homolog protein 2; LOW QUALITY PROTEIN: homeobox even-skipped homolog protein 2
Gene Symbols EVX2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of rat Evx2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288155
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.