Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EWSR1 Antibody (5H7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192686F
Description
EWSR1 Monoclonal specifically detects EWSR1 in Human, Mouse, Rat, Canine, Equine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
EWSR1 | |
Monoclonal | |
1mg/mL | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
bK984G1.4, Ewing sarcoma breakpoint region 1, Ewing sarcoma breakpoint region 1 protein, Ewings sarcoma EWS-Fli1 (type 1) oncogene, EWS oncogene, EWSRNA-binding protein EWS | |
Mouse | |
85 kDa | |
0.1 mL | |
Cancer, Cellular Markers, Neuroscience, Tumor Suppressors | |
2130 | |
Store at 4C in the dark. | |
IgG2b |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
5H7 | |
FITC | |
Q01844 | |
EWSR1 | |
Full length recombinant human EWSR1 expressed in and purified from E. coli. [UniProt# Q01844] | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Equine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction