Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EWSR1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB200183AF488
Description
EWSR1 Polyclonal antibody specifically detects EWSR1 in Human samples. It is validated for Western BlotSpecifications
EWSR1 | |
Polyclonal | |
Western Blot | |
bK984G1.4, Ewing sarcoma breakpoint region 1, Ewing sarcoma breakpoint region 1 protein, Ewings sarcoma EWS-Fli1 (type 1) oncogene, EWS oncogene, EWSRNA-binding protein EWS | |
A synthetic peptide that maps to a region between residues 600 and the C-terminus (residue 656) of human Ewing sarcoma breakpoint region 1 using the numbering given in SwissProt entry Q01844 (GeneID 2130). | |
0.1 mL | |
Cancer, Cellular Markers, Neuroscience, Tumor Suppressors | |
2130 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction