Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EWSR1 Recombinant Rabbit Monoclonal Antibody (9G3H1)
GREENER_CHOICE

Catalog No. PIMA535826
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535826 Supplier Invitrogen™ Supplier No. MA535826
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MASTDYSTYS QAAAQQGYSA YTAQPTQGYA QTTQAYGQQS YGTYGQPTDV SYTQAQTTAT YGQTAYATSY GQPPTGYTTP TAPQAYSQPV QGYGTGAYDT TTATVTTTQA SYAAQSAYGT QPAYPAYGQQ PAATAPTRPQ DGNKPTETSQ PQSSTGGYNQ PSLGYGQSNY SYPQVPGSYP MQPVTAPPS.

This gene encodes a multifunctional protein that is involved in various cellular processes, including gene expression, cell signaling, and RNA processing and transport. The protein includes an N-terminal transcriptional activation domain and a C-terminal RNA-binding domain. Chromosomal translocations between this gene and various genes encoding transcription factors result in the production of chimeric proteins that are involved in tumorigenesis. These chimeric proteins usually consist of the N-terminal transcriptional activation domain of this protein fused to the C-terminal DNA-binding domain of the transcription factor protein. Mutations in this gene, specifically a ttranslocation, are known to cause Ewing sarcoma as well as neuroectodermal and various other tumors. Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 1 and 14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EWSR1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 9G3H1
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EWSR1
Gene Accession No. Q01844, Q61545
Gene Alias AC002059.7; bK984G1.4; Ewing sarcoma breakpoint region 1; Ewing sarcoma breakpoint region 1 protein; Ewing sarcoma homolog; Ewings sarcoma EWS-Fli1 (type 1) oncogene; Ews; EWS oncogene; EWS RNA binding protein 1; EWS RNA-binding protein 1; EWS RNA-binding protein variant 6; EWS-FLI1; Ewsh; EWSR1; RNA-binding protein EWS
Gene Symbols EWSR1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14030, 2130, 289752
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.