Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ex 26
GREENER_CHOICESDP

Catalog No. 583350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-335-0 10 mg
58-331-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 58-335-0 Supplier Tocris Bioscience™ Supplier No. 5833/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective S1P1 receptor antagonist

Ex 26 is a potent and selective S1P1 receptor antagonist (IC50 = 0.93 nM). Exhibits > 3,000-fold selectivity for S1P1 over S1P2, S1P3, S1P4 and S1P5. Induces lymphocyte sequestration in peripheral lymph nodes and upregulates S1P1 receptor expression. Reduces disease severity in mice with EAE. Increases IFN-α production in stimulated dendritic cells in vivo.

Specifications

Chemical Name or Material 1-[[[5'-[[1-(4-Chloro-3-methylphenyl)ethyl]amino]-2'-fluoro-3,5-dimethyl[1,1'-biphenyl]-4-yl]carbonyl]amino]-cyclopropanecarboxylic acid
CAS 144678-63-7
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Molecular Formula C28H28ClFN2O3
Formula Weight 494.98
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.