Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ EX 527
GREENER_CHOICESDP

Catalog No. 278050 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-805-0 50 mg
27-801 1 mg
27-801-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 27-805-0 Supplier R&D Systems™ Supplier No. 2780/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective SIRT1 inhibitor

EX 527 is a selective inhibitor of SIRT1 that does not inhibit histone deacetylase (HDAC) or other sirtuin deacetylase family members (IC50 values are 98, 19600, 48700, > 100000 and > 100000 nM for SIRT1, SIRT2, SIRT3, HDAC and NADase respectively). In an animal model of Huntington's disease, EX 527 rescues neuronal degeneration, extends lifespan, reduces huntingtin inclusions and prevents ventricular enlargement. EX 527 enhances p53 acetylation in response to DNA damaging agents. Orally bioavailable and brain penetrant.

Chemical Identifiers

CAS 49843-98-3
Molecular Formula C13H13ClN2O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 248.71
Purity >98%
Molecular Weight (g/mol) 248.71
Chemical Name or Material EX 527
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.