Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva ExcelGel SDS Buffer Strips

Catalog No. 45000246 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
6 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-246 6 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-246 Supplier Cytiva Supplier No. 17134201

missing translation for 'validMainframeMsg'

Semi dry precast buffer strips for horizontal SDS electrophoresis. Cytiva Lifescience™ ExcelGel SDS Buffer Strips are designed for use with ExcelGel Precast Gels.

  • Designed for use with ExcelGel Precast Gels for SDS-PAGE
  • Include strips for both anode and cathode

TRUSTED_SUSTAINABILITY

Specifications

Formulation Acrylamide 0.55%
For Use With (Application) SDS-PAGE
Quantity 6 Pack
Storage Requirements Cool, dry place
Product Type ExcelGel SDS Buffer strips
For Use With (Equipment) Multiphor II Flatbed Electrophoresis System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.