Learn More
Excelta™ Excelta™ Acetal Replacement Tips for Extreme Precision 169C-RTW Tweezers
Shop All Excelta Corporation ProductsDescription
- Includes two tips and three screws
- Resistant to chemicals and solvents
- For cleanroom use
- Tip size: 2 x 1.13mm (0.08 x 0.045 in.)
Specifications
Specifications
| Dimensions (L x W) | 2 x 1.13 mm (0.08 x 0.045 in.) |