Learn More
Excelta™ Excelta™ Carbofib™ Extreme Precision Replacement Tips for 169E-RT Tweezers
Catalog No. 18100940 Shop All Excelta Corporation Products
Change view
Click to view available options
Replacement tips for 169E-RT tweezers
Catalog No.
18-100-940 Replacement tips for 169E-RT tweezers
1 options
Catalog No. 18-100-940 Supplier Excelta™ Supplier No. 169ERTX
Set of replacement tips for 169E-RT tweezers
  • Includes two tips and three screws
  • Tip size: 8 x 0.8mm (0.12 x 0.032 in.)
Specifications
Dimensions (L x W) 8 x 0.8 mm (0.12 x 0.032 in.)