Learn More
Excelta™ Excelta™ Carbofib Replacement Tips for 769 Series High Precision Tweezers
Catalog No. 18100945 Shop All Excelta Corporation Products
Catalog No. 18-100-945 Supplier Excelta™ Supplier No. 769DRTX
Replaceable tips for extreme precision Carbofib 769 tweezers
  • Includes set of two tips and four screws
  • Made of PTFE based material with 30% carbon fibers
  • Working temp is 249°F
  • 769-RTX replacement tips for 769-RT
  • Tip Size: 0.5mm (0.20 in.)
  • 769D-RTX replacement tips for 769D-RT
  • Tip Size: 1.38mm (0.055 in.)