Learn More
Excelta™ Excelta™ Carbofib Extreme Precision Replacement Tips for 162D-RT Tweezers
Catalog No. 18100924 Shop All Excelta Corporation Products
Change view
Click to view available options
Replacement tips for 169-RT tweezers
Catalog No.
18-100-924 Replacement tips for 169-RT tweezers
1 options
Catalog No. 18-100-924 Supplier Excelta™ Supplier No. 169RTX
Set of replacement tips for 162D-RT tweezers
  • Includes two tips and three screws
  • Extreme Precision (Four star rating)
  • Working temperature of Carbofib is 249degreeF
  • Tool is conductive (102 ohms/sq)
  • Tip size: 0.3 x 0.23mm (0.012 x 0.009 in.)
Specifications
Product Type Replacement Tips for 169-RT tweezers
Material Carbofib
Dimensions (L x W) Tip: 0.3 x 0.23 mm (0.012 x 0.009 in.)