Learn More
Excelta™ Excelta™ Extreme Precision Stainless Steel Tweezer with Straight Soft Replaceable Carbofib Tips
Catalog No. 18100939 Shop All Excelta Corporation Products
Catalog No. 18-100-939 Supplier Excelta™ Supplier No. 169ERT
Carbofib has better rigidity, tensile and flexural strength with fatigue and creep resistance
  • ESD safe conductive tool (102 ohms/sq))
  • Extreme Precision: Four star
  • Working temperature of Carbofib: 249°F
  • Available in various tip widths