Learn More
Excelta™ Excelta™ Extreme Precision Stainless Steel Tweezers with Straight Soft Replaceable Acetal Tips
Catalog No. 18100936 Shop All Excelta Corporation Products
Catalog No. 18-100-936 Supplier Excelta™ Supplier No. 169DRTW
Acetal tips offers resistance to chemicals and solvents
  • For cleanroom use
  • Extreme Precision: Four star
  • Overall length: 125mm (5 in.)
Specifications
Length (Metric) 125 mm
Tip Style Straight