Learn More
Excelta™ High Precision 159 Series Reverse Action Soft Replaceable Tip 5 in. Tweezers
Catalog No. 18100919 Shop All Excelta Corporation Products
Catalog No. 18-100-919 Supplier Excelta™ Supplier No. 159RTN
Add to Cart
Add to Cart
Features high rigidity and flFeatures high rigidity and flexural strengthexural strength
  • Tips are made of polymer based material with 30% carbon fibers (Carbofib)
  • Remain closed at rest
  • High Precision: Three star
  • Overall length: 125mm (5 in.)
  • 159-RTN-With 0.3mm (.012 in.) wide tips
  • 159D-RTN-With 1.5mm (0.060 in.) wide tips