Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Excelta™ High-Precision ESD Safe Brushes
Bristles are mechanically clamped into handle for maximum fiber retention without use of adhesives that could contaminate work areas
Supplier: Excelta™ 196ESD
Description
- Stainless-steel or plastic handles
- Horse hair bristles
Specifications
Cleaning Brush | |
Soft | |
Flat | |
1.5 in. | |
3.8 cm |
Horse Hair | |
Plastic | |
5.75 in. | |
14.6 cm | |
Fine |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction