Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Blue Micro Spatulas, plastic handle

Catalog No. 17456036 Shop All Excelta Corporation Products
Change view
Click to view available options
Length (Metric):
6.4 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
17-456-036 6.4 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-456-036 Supplier Excelta™ Supplier No. 260B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Can be used to apply lubricants to small moving parts

High precision

Specifications

Color Blue
Product Type Micro Spatula
For Use With (Application) Used to apply lubricants to small moving parts and for applying adhesives during electronic
Length (English) 2.5 in.
Precision High
Includes Plastic handle
Handle Material Plastic
Material Stainless Steel (440)
Length (Metric) 6.4 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.