Learn More
Excelta™ Roto-Pic Vacuum System Vacuum Cups
Catalog No. 17468550 Shop All Excelta Corporation Products
Change view
Click to view available options
Product Type:
Roto-pic Circovac Vacuum Cup
Roto-pic Vacuum Cup
Roto-pic Verto-Vac Vacuum Cup
Diameter (Metric) Inside:
0.6 cm
1 cm
1.2 cm
Not Applicable
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Diameter (Metric) Inside
17-468-550 Roto-pic Vacuum Cup 0.6 cm
17-468-548 Roto-pic Vacuum Cup 1 cm
17-468-549 Roto-pic Vacuum Cup 1.2 cm
17-468-552 Roto-pic Verto-Vac Vacuum Cup Not Applicable
17-468-551 Roto-pic Circovac Vacuum Cup Not Applicable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-468-550 Supplier Excelta™ Supplier No. 2052
Add to Cart
Add to Cart
High temperature silcone rubber with brass fittings.
  • High temperature silicone rubber
  • Includes brass fittings
Specifications
Product Type Roto-pic Vacuum Cup
For Use With (Equipment) Hand piece and probes
Material Silicone Rubber
Diameter (English) Inside 0.25 in.
Diameter (Metric) Inside 0.6 cm
Includes Brass fittings