Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Shear Carbon Steel Cutter

Catalog No. 18100950 Shop All Excelta Corporation Products
Change view
Click to view available options
Length (English):
4.25 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
18-100-950 4.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-100-950 Supplier Excelta™ Supplier No. SE25

The Shear Carbon Steel Cutter reduces both cutting effort and mechanical shock transmitted to component.

  • The SE-25 small micro shear with tapered head is lightweight and easy to handle.
  • Shear cutting action reduces both cutting effort and mechanical shock transmitted to the component.

Specifications

Product Type Shear Cutter
Blade Material Carbon Tool Steel
Grade General Precision
Head Style Tapered
Length (English) 4.25 in.
Length (Metric) 106.5 mm
Material Carbon Steel
Size Medium
Cut Type Shear
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.