Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EXD1 Antibody (OTI5B5), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272443MFV500
Description
EXD1 Monoclonal antibody specifically detects EXD1 in Human samples. It is validated for Flow CytometrySpecifications
EXD1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
161829 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI5B5 | |
Flow Cytometry | |
EXDL1, exonuclease 3'-5' domain containing 1, exonuclease 3'-5' domain-containing protein 1, exonuclease 3'-5' domain-like 1, Exonuclease 3'-5' domain-like-containing protein 1, MGC33637 | |
Full length human recombinant protein of human EXD1 (NP_689809) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction