Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EXD1 Antibody (OTI5B5), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272443PE
Description
EXD1 Monoclonal specifically detects EXD1 in Human samples. It is validated for Flow Cytometry.Specifications
EXD1 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
EXD1 | |
Full length recombinant protein of human EXD1 (NP_689809) produced in HEK293T cells. | |
0.1 mL | |
161829.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI5B5 | |
Flow Cytometry | |
EXDL1, exonuclease 3′-5′ domain containing 1, exonuclease 3′-5′ domain-containing protein 1, exonuclease 3′-5′ domain-like 1, Exonuclease 3′-5′ domain-like-containing protein 1, MGC33637 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction