Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXD1 Monoclonal Antibody (OTI5B5), TrueMAB™, OriGene
SDP

Catalog No. 502541794
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1794 30 μL
50-167-3182 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1794 Supplier OriGene Supplier No. TA502098S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EXD1 Monoclonal antibody specifically detects EXD1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen EXD1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI5B5
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CNDP1
Gene Accession No. Q96KN2
Gene Alias 4932702D22Rik; Exd1; EXDL1; exonuclease 3'-5' domain containing 1; exonuclease 3'-5' domain-containing protein 1; exonuclease 3'-5' domain-containing protein 1; piRNA biogenesis protein EXD1; exonuclease 3'-5' domain-like 1; exonuclease 3'-5' domain-like-containing protein 1; Inactive exonuclease EXD1; mExd1; piRNA biogenesis protein EXD1; RGD1561009; Vexonuclease 3'-5' domain-like 1
Host Species Mouse
Immunogen Full length human recombinant protein of human CNDP1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84735
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.