Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exel International Disposable Spinal Needles
Click to view available options
Needle Gauge:
18 G
20 G
22 G
25 G
Color:
Black
Orange
Pink
Yellow
Description
- Latex-free
- Sterile
- Nontoxic
- Nonpyrogenic
- Convenient blister package
- Siliconized for smooth, comfortable penetration
Specifications
Specifications
| Color | Pink |
| Needle Gauge | 18 G |
| For Use With (Application) | Siliconized for smooth, comfortable penetration |
| Latex Free | Yes |
| Length (English) Needle | 3.5 in. |
| Product Type | Spinal Needle |
| Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction