Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exendin-4 Antibody (20) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Exendin-4 |
---|---|
Clone | 20 |
Concentration | 1 mg/mL |
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019342
![]() |
Novus Biologicals
NBP2235770.025ML |
0.025 mL |
Each for $206.50
|
|
|||||
NBP22357701
![]() |
Novus Biologicals
NBP2235770.1ML |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Exendin-4 Monoclonal specifically detects Exendin-4 in Reptile samples. It is validated for ELISA.Specifications
Exendin-4 | |
1 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
Synthetic exendin-4 adsorbed onto aluminum hydroxide gel | |
Primary | |
Store at 4C. Do not freeze. |
20 | |
ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Human | |
Exendin4 | |
IgG1 κ | |
Protein A or G purified | |
Specific for exendin-4. No cross-reactivity with GLP-1, GLP-2 (human) or glucagon coated on ELISA wells |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title