Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Exendin-4 Antibody (20), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP223577MFV500
Description
Exendin-4 Monoclonal antibody specifically detects Exendin-4 in Reptile samples. It is validated for ELISASpecifications
Exendin-4 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Reptile | |
Purified |
ELISA | |
20 | |
ELISA | |
Exendin4 | |
Synthetic exendin-4 | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction