Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exendin-4 Antibody (24), Alexa Fluor™ 350, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP223579AF350

Catalog No. NP22379A30


Only null left
Add to Cart

Description

Description

Exendin-4 Monoclonal specifically detects Exendin-4 in Reptile samples. It is validated for ELISA.
Specifications

Specifications

Exendin-4
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
Mouse
0.1 mL
Specific for exendin-4. No cross-reactivity with GLP-1, GLP-2 (human) or glucagon coated on ELISA wells.
Store at 4°C in the dark.
ELISA
24
ELISA
Exendin4
Synthetic exendin-4 adsorbed onto aluminum hydroxide gel
Primary
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.