Learn More
Exendin-4 Antibody (24) - BSA Free, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP2235790.1ML
 View more versions of this product
Catalog No. NBP22357901
Only null left
Add to Cart
Description
Description
Exendin-4 Monoclonal specifically detects Exendin-4 in Reptile samples. It is validated for ELISA.
Specifications
Specifications
Exendin-4
Monoclonal
1 mg/mL
ELISA 1:100-1:2000
Exendin4
Synthetic exendin-4 adsorbed onto aluminum hydroxide gel
0.1 mL
Specific for exendin-4. No cross-reactivity with GLP-1, GLP-2 (human) or glucagon coated on ELISA wells.
Store at 4C. Do not freeze.
IgG1 κ
ELISA
24
Unconjugated
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Mouse
Protein A or G purified
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only