Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exendin 4 Monoclonal Antibody (20)
GREENER_CHOICE

Catalog No. ABS0122002
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS0122002 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS0122002 Supplier Thermo Scientific Supplier No. ABS0122002
Add to Cart
Add to Cart

Exendin 4 Monoclonal Antibody (20)

Exendin 4 Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Exendin 4
Applications ELISA
Classification Monoclonal
Clone 20
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene Alias Exenatide
Host Species Mouse
Immunogen synthetic exendin-4.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.