Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Exendin 4 Monoclonal Antibody (24)
GREENER_CHOICE

Catalog No. ABS0122402
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS0122402 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS0122402 Supplier Invitrogen™ Supplier No. ABS0122402
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABS 012-24-02 detects exendin-4 from human samples. This antibody does not cross react with GLP-1, GLP-2 or glucagon. ABS 012-24-02 has been successfully used in ELISA procedures. ABS 012-24-02 binds to exendin-4 when coated on ELISA wells and reacts specifically with exendin-4 in solution giving a Ka of 1.8x10E8 in an inhibition ELISA. The binding between ABS 012-24-02 and exendin-4 is disrupted by 0.1 M glycin pH 3. There is full reaction with exendin-4 (9-39) when ABS 012-24-02 is used as coat in ELISA showing an epitope in the 9 - 39 region.

Exendin-4 is a 39-amino-acid peptide found in venom from the Gila monster Helicoderma suspectum. It is a member of the glucagon-secretin family of peptide hormones and neuropeptides. Exendin-4 is a potent agonist of the GLP-1 receptor and hence a potent stimulator of insulin secretion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Exendin 4
Applications ELISA
Classification Monoclonal
Clone 24
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene Alias Exenatide
Host Species Mouse
Immunogen Carrier-coupled exendin-4.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.