Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXO1 Monoclonal Antibody (266), Invitrogen™

Catalog No. PIMA512262
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512262 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512262 Supplier Thermo Scientific Supplier No. MA512262

Mouse Monoclonal Antibody

MA5-12262 targets Exo1 in WB applications and shows reactivity with Human and mouse samples. The MA5-12262 immunogen is recombinant full length mouse Exo1 protein.

Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. Required for muscle atrophy following denervation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXO1
Applications Western Blot
Classification Monoclonal
Clone 266
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene EXO1
Gene Accession No. Q9QZ11, Q9UQ84
Gene Alias 5730442G03Rik; Exo1; EXOI; Exonuclease 1; exonuclease I; HEX1; hExo1; hExoI; mExo1; Msa; rad2 nuclease family member, homolog of S. cerevisiae exonuclease 1
Gene Symbols EXO1
Host Species Mouse
Immunogen Recombinant full length mouse Exo1 protein
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26909, 9156
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.