Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558071 Supplier Invitrogen™ Supplier No. PA558071
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FKLQQHQSMP GTMAEAEDLD GGTLSRQHNC GTPLPVSSEK DMIRQMMIKI FRCIEPELNN LIALGDKIDS FNSLYMLVKM SHHVWTA Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.

The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of the exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. Alternatively spliced transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EXOC1
Gene Accession No. Q9NV70
Gene Alias 2810407P21Rik; A730011E05Rik; BM-012; BM-102; Exoc1; Exocyst complex component 1; exocyst complex component Sec3; Sec3; SEC3L1; SEC3-like 1; Sec3p
Gene Symbols EXOC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EXOC1. Recombinant protein control fragment (Product #RP-96615).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55763
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.