Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EXOC2 Antibody (SR1032), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322632100UL
Description
EXOC2 Monoclonal antibody specifically detects EXOC2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
EXOC2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1032 | |
Western Blot 1:500-1:2000 | |
exocyst complex component 2, Exocyst complex component Sec5, FLJ11026, SEC5, SEC5L1, SEC5-like 1, SEC5-like 1 (S. cerevisiae), Sec5p | |
A synthesized peptide derived from human EXOC2 (Uniprot #: Q96KP1) | |
100 μg | |
Signal Transduction | |
55770 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction