Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EXOC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546730
Description
Peptide sequence: MLDLDNDTRP SVLGHLSQTA SLKRGSSFQS GRDDTWRYKT PHRVAFVEKL Sequence homology: Cow: 85%; Dog: 100%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and the functions of the exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. This interaction has been shown to mediate filopodia formation in fibroblasts.
Specifications
| EXOC2 | |
| Polyclonal | |
| Unconjugated | |
| Exoc2 | |
| 2410030I24Rik; AI648199; Exoc2; Exocyst complex component 2; Exocyst complex component 2-like protein; exocyst complex component Sec5; rSec5; Sec5; SEC5 homolog; Sec5 protein; Sec5l1; SEC5-like 1; Sec5p | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55770 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96KP1 | |
| Exoc2 | |
| Synthetic peptide directed towards the N-terminal of human EXOC2 (aa 406-455). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction