Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOC3 Monoclonal Antibody (9H5)
GREENER_CHOICE

Catalog No. MA125480
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA125480 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA125480 Supplier Invitrogen™ Supplier No. MA125480
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Western blot detects a band at ~87 kDa.

The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOC3
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.1mM PMSF and no preservative; pH 7.2
Gene EXOC3
Gene Accession No. O60645, Q0V8C2, Q62825, Q6KAR6
Gene Alias 2810050O03Rik; E430013E20Rik; EXOC3; EXOC3L; EXOC3L1; Exocyst complex component 3; exocyst complex component 3 like 1; exocyst complex component 3-like 1; exocyst complex component 3-like protein; Exocyst complex component Sec6; I79_024581; KIAA0895L; KIAA0895-like ortholog; LIM/homeobox protein Lhx2 pseudogene; protein Jiangli; RGD1311132; rSec6; Sec 6 homolog; Sec6; Sec6 protein; SEC6L1; SEC6-like 1; Sec6p
Gene Symbols EXOC3
Host Species Mouse
Immunogen Recombinant rat Sec6.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100356252, 100624948, 100756215, 101103266, 11336, 211446, 252881, 478633, 513138
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Sheep, Hamster, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.