Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOC3L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556196
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556196 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556196 Supplier Invitrogen™ Supplier No. PA556196
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGSLELGPEA DVSQLEPLLT LENIEQLEAT FVANIQASVS QWLQNALDGE VAEWGREHGP NTDPSGSYYS PMPAIVLQIL EENIRVASLV SESLQQRVHG MALSELGTFL RSFSDALIRF SRDHFRGKSM APHYVPYL Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 78%.

As part of the exocyst, may play a role in regulated exocytosis of insulin granules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOC3L1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EXOC3L1
Gene Accession No. Q86VI1
Gene Alias C730015A04Rik; EXOC3L; EXOC3L1; exocyst complex component 3 like 1; exocyst complex component 3-like; exocyst complex component 3-like 1; exocyst complex component 3-like protein; Protein Jiangli
Gene Symbols EXOC3L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EXOC3L1. Recombinant protein control fragment (Product #RP-92438).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283849
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.