Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOC3L4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560455
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560455 Supplier Invitrogen™ Supplier No. PA560455
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQALNDGPAT GHSQATPEVP SGVMNGVSQQ ASTGAASEEL KPEAEGKSVA DLITERQLLA AFEQLLRLET LLVAEKASRT FEQDPTAFAR RAMDVCLLYD GLA Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 67%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOC3L4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EXOC3L4
Gene Accession No. Q17RC7
Gene Alias C14orf73; EXOC3L4; exocyst complex component 3 like 4; exocyst complex component 3-like 4; exocyst complex component 3-like protein 4; SEC6-like protein C14orf73
Gene Symbols EXOC3L4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EXOC3L4. Recombinant protein control fragment (Product #RP-100319).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91828
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.