Learn More
EXOC4 Mouse anti-Human, Clone: OTI1E9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811242

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
EXOC4 | |
Monoclonal | |
Unconjugated | |
EXOC4 | |
SEC8, SEC8L1, Sec8p | |
Mouse | |
Affinity Chromatography | |
RUO | |
60412 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1E9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96A65 | |
EXOC4 | |
Full length human recombit protein of human EXOC4 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.